Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boraxcarmine powder, TCI America™
SDP

Catalog No. B05165G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B05165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B05165G Supplier TCI America Supplier No. B05165G
Add to Cart
Add to Cart

Chemical Identifiers

MDL Number MFCD01310220

Specifications

Color Purple-Red
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material Boraxcarmine powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.