Learn More
Analytical Sales & Services Border Gasket for 384-Well Glass Plate
Catalog No. 5000804248
Encompass
Border Gasket for 384-Well Glass Plate\n\n• For Nano Nest Parallel Synthesis and Photoredox Blocks that use Glass Collection Plates
Catalog No. 50-008-04248 Supplier Analytical Sales & Services Supplier No. 1626003
Border Gasket for 384-Well Glass Plate\n\n• For Nano Nest Parallel Synthesis and Photoredox Blocks that use Glass Collection Plates