Learn More
HIMEDIA Bordet Gengou Agar Base, M175, Microbiology Culture Media, Dehydrated Culture Media, Agar Base, Bordetella pertussis Isolation, Requires Blood, Powder, 500 g
Catalog No. NC4013702
Encompass_Preferred
Bordet Gengou Agar Base is the classic medium for detecting and isolating Bordetella pertussis and Bordetella parapertussis, the agents of whooping cough. Potato infusion and peptone supply nutrition while a high agar level gives a firm surface at final pH 6.7. Prepared with added glycerol and defibrinated blood, it supports these highly fastidious organisms and reveals their characteristic hemolysis. A reference base for clinical and respiratory microbiology. Dehydrated powder, store in a cool dry place. Packaged as 500 g.
Catalog No. NC4013702 Supplier HIMEDIA Supplier No. M175500G
Bordet Gengou Agar Base is the classic medium for detecting and isolating Bordetella pertussis and Bordetella parapertussis, the agents of whooping cough. Potato infusion and peptone supply nutrition while a high agar level gives a firm surface at final pH 6.7. Prepared with added glycerol and defibrinated blood, it supports these highly fastidious organisms and reveals their characteristic hemolysis. A reference base for clinical and respiratory microbiology. Dehydrated powder, store in a cool dry place. Packaged as 500 g.