Learn More
ATCC Bordetella bronchiseptica
Catalog No. 502381975
Encompass
Type strain. Produces restriction endonuclease BbrI.
Catalog No. 50-238-1975 Supplier ATCC Supplier No. 19395
Type strain. Produces restriction endonuclease BbrI.