Learn More
ATCC Bordetella bronchiseptica
Supplier:  ATCC 4617
Encompass
Bordetella bronchiseptica strain NRRL B-140 is a whole-genome sequenced strain with applications in susceptibility disc testing for colistin and polymyxin B.
Catalog No. 50-238-3739