Learn More
ZeptoMetrix Bordetella Bronchiseptica Z339, Titered
SDP
Catalog No. 22032222 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
22-032-222 1 mL
1 options
Catalog No. 22-032-222 Supplier ZeptoMetrix Supplier No. 0804312
Add to Cart
Add to Cart
Live, titered Fusobacterium nucleatum for diagnostic assay testing and development.
  • Suitable for determining limit of detection (LOD)
  • Supports diagnostic assay development and cross-reactivity studies
  • Can be used as a control for nucleic acid tests
  • When used as a nucleic acid test control, the same amplification protocols used for clinical specimens should be followed
Specifications
Product Type Bacteria Control
Sterility Non-sterile
Quantity 1 mL