Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vela Diagnostics Great Basin Bordetella Direct Test

Catalog No. 23052003
Change view
Click to view available options
Quantity:
10 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-052-003 10 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-052-003 Supplier Vela Diagnostics Supplier No. 300782
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Offers cost-effective, STAT testing without paying for unnecessary analytes, providing definitive results in under 2 hours allowing prompt treatment/reporting, and has been validated with several media types, making it compatible with virtually any nasopharyngeal specimen collection protocol.

This syndromic panel simultaneously detects and differentiates specific gene targets from the most prevalent bacterial causes of acute gastroenteritis – Salmonella, Shigella, C. jejuni, C. coli, Shiga toxin-producing E. coli (stx1, and stx2), and E. coli serotype O157 (if Shiga toxin detected).

Specifications

Content And Storage 2°C to 8°C
Description Sample-to-result test: Bordetella pertussis
Clia Complexity Moderate
Detectable Analytes Bordatella
Sample Type Nasopharyngeal Swab
Quantity 10 Assays
Sample Size 50 μL
Test Time ≈90 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.