Learn More
ATCC Bordetella holmesii
Catalog No. 502380566
Encompass
Bordetella holmesii strain CIP 104395 was isolated from the dog bit wound of a human in California. This bacterial strain is grown in a 5% CO2 atmosphere on heart infusion medium with 5% rabbit blood.
Catalog No. 50-238-0566 Supplier ATCC Supplier No. 700052
Bordetella holmesii strain CIP 104395 was isolated from the dog bit wound of a human in California. This bacterial strain is grown in a 5% CO2 atmosphere on heart infusion medium with 5% rabbit blood.