Learn More
ATCC Bordetella parapertussis; 522; genomic DNA
Catalog No. 502383256
Encompass
Genomic DNA isolated from Bordetella parapertussis strain NCTC 5952 [522] (ATCC 15311). This whole-genome sequenced preparation can be used in PCR and other molecular biology applications.
Catalog No. 50-238-3256 Supplier ATCC Supplier No. 15311D5
Genomic DNA isolated from Bordetella parapertussis strain NCTC 5952 [522] (ATCC 15311). This whole-genome sequenced preparation can be used in PCR and other molecular biology applications.