Learn More
ATCC Bordetella pertussis; 18323
Catalog No. 502384339
Encompass
Bordetella pertussis strain 18323 [NCTC 10739] is a whole-genome sequenced bacterial type strain that has applications in testing the potency of the pertussin vaccine.
Catalog No. 50-238-4339 Supplier ATCC Supplier No. 9797
Bordetella pertussis strain 18323 [NCTC 10739] is a whole-genome sequenced bacterial type strain that has applications in testing the potency of the pertussin vaccine.