Learn More
ATCC Mycobacterium; talmoniae sp. nov.; Strain: NE-TNMC-100812
Catalog No. 502380483
Encompass
Type strain. Resistant to ciprofloxacin, linezolid, ethambutol, and bactrim. Slow growing strain.
Catalog No. 50-238-0483 Supplier ATCC Supplier No. BAA2683
Type strain. Resistant to ciprofloxacin, linezolid, ethambutol, and bactrim. Slow growing strain.