Learn More
ATCC Bordetella pertussis; H792
Catalog No. 502379892
Encompass
The depositor states that this strain doesn't produce pertactin and that there is a STOP 1273 nucleotide mutation.
Catalog No. 50-237-9892 Supplier ATCC Supplier No. BAA2701
The depositor states that this strain doesn't produce pertactin and that there is a STOP 1273 nucleotide mutation.