Learn More
ATCC Bordetella pertussis; Strain: H762
Catalog No. 502384678
Encompass
Bordetella pertussis strain H762 is a bacterium that was isolated in 2011 from a human in the United States. The depositor states that this culture is pertactin negative.
Catalog No. 50-238-4678 Supplier ATCC Supplier No. BAA3205
Bordetella pertussis strain H762 is a bacterium that was isolated in 2011 from a human in the United States. The depositor states that this culture is pertactin negative.