Learn More
Creative Biomart Recombinant Polyvalent Dengue Antigen, His-tagged
Catalog No. 502084847
Encompass_Preferred
Polyvalent dengue antigens are 18 kDa proteins containing subtype 1,2,3 and 4 and each of them has equal amount protein. They are a new group of antigens specifically designed for ELISA assay. All the
Catalog No. 50-208-4847 Supplier Creative Biomart Supplier No. VAC0007
Polyvalent dengue antigens are 18 kDa proteins containing subtype 1,2,3 and 4 and each of them has equal amount protein. They are a new group of antigens specifically designed for ELISA assay. All the