Learn More
Grainger Bore Tank Valve Large 1-19/32 1/4 NPT
Catalog No. 502750703
Encompass_Preferred
The Haltec Bore Tank Valve is a 1/2" NPT brass ball valve designed for use in bore tanks. It has a maximum pressure rating of 300 PSI and temperature rating of 250°F. The valve features a chrome-plated brass ball for durability, blowout-proof stem, and adjustable packing gland. It can be used to control flow and shut off water supply in residential and commercial bore tank applications. The valve body and ball are made of brass for corrosion resistance.
Catalog No. 50-275-0703 Supplier Grainger Supplier No. 140Y20
The Haltec Bore Tank Valve is a 1/2" NPT brass ball valve designed for use in bore tanks. It has a maximum pressure rating of 300 PSI and temperature rating of 250°F. The valve features a chrome-plated brass ball for durability, blowout-proof stem, and adjustable packing gland. It can be used to control flow and shut off water supply in residential and commercial bore tank applications. The valve body and ball are made of brass for corrosion resistance.