Learn More
Biocare Medical, LLC Borg Decloaker, RTU - 1000 ml
SDP
Supplier:  Biocare Medical, LLC BD1000MM
Encompass_Preferred
Reveal Decloaker is a heat retrieval solution that is suitable for most antibody-antigen staining and reduces non-specific background staining, blocks endogenous peroxidase and removes mercury crystals. Reveal Decloaker can be used with Biocare's digital electric pressure cooker (Decloaking Chamber), a steamer, a water bath, or a microwave oven. For many antibodies, titers are doubled and background staining is significantly reduced when compared to citrate buffer, pH 6.0. Reveal Decloaker incorporates AssureTM technology, a color-coded high temperature pH indicator solution. The end-user is assured by visual inspection that the solution is at the correct dilution and pH. This product is specially formulated for superior pH stability at high temperatures and will help prevent the possibility of losing pH sensitive antigens. Reveal Decloaker is non-toxic, non-flammable, odorless and sodium azide and thimerosal free.
Catalog No. 50-823-71