Learn More
Alkali Scientific Boric Acid, 2.5 Kilograms
SDP
Catalog No. 5000660218
Encompass_Preferred
Boric Acid is a laboratory-grade reagent provided in a crystalline form with a weight of 2.5 kg. It is commonly used in various chemical processes, including pH buffering, antimicrobial formulations, and as a flux in metal welding. Boric acid serves as an effective pH stabilizer, particularly in acidic solutions, and is widely used in the preparation of borate buffers for industrial and laboratory applications. The crystalline form is highly soluble in water, making it easy to use in aqueous solutions. In laboratory environments, it plays a critical role in assays, chemical syntheses, and as a preservative in certain chemical formulations.
Catalog No. 50-006-60218 Supplier Alkali Scientific Supplier No. BOR006CP
Boric Acid is a laboratory-grade reagent provided in a crystalline form with a weight of 2.5 kg. It is commonly used in various chemical processes, including pH buffering, antimicrobial formulations, and as a flux in metal welding. Boric acid serves as an effective pH stabilizer, particularly in acidic solutions, and is widely used in the preparation of borate buffers for industrial and laboratory applications. The crystalline form is highly soluble in water, making it easy to use in aqueous solutions. In laboratory environments, it plays a critical role in assays, chemical syntheses, and as a preservative in certain chemical formulations.