Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Acid, 2% w/v with Methyl Red-Bromocresol Green Indicators, Reagents

Catalog No. 01336574
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-574 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-574 Supplier Reagents Holdings Llc Supplier No. C71042204A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boric Acid, 2% w/v with Methyl Red-Bromocresol Green Indicators, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Boric Acid, 2% w/v with Methyl Red-Bromocresol Green Indicators
pH 4.5 to 5
Color Purple
CAS 7732-18-5, 10043-35-3
Boiling Point 100°C
Packaging Poly Bottle
Specific Gravity 1.0074
Quantity 4 L
Physical Form Liquid
Percent Purity 2%
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.