Learn More
Strem, An Ascensus Company CAS# 10043-35-3. 2kg. Boric acid, min. 99.5%. MFCD00011337
SDP
Catalog No. 5090113708
Encompass
CAS# 10043-35-3. 2kg. Boric acid, min. 99.5%. MFCD00011337. Molecular Weight: 61.83. Molecular Formula: H3BO3. Color/form: white xtl. Strem# 93-0504. http://www.strem.com/catalog/v/93-0504/
Catalog No. 50-901-13708 Supplier Strem, An Ascensus Company Supplier No. 9305042000G
CAS# 10043-35-3. 2kg. Boric acid, min. 99.5%. MFCD00011337. Molecular Weight: 61.83. Molecular Formula: H3BO3. Color/form: white xtl. Strem# 93-0504. http://www.strem.com/catalog/v/93-0504/