Learn More
Strem, An Ascensus Company CAS# 10043-35-3. 10g. Boric acid (99.9995%-B) PURATREM. MFCD00011337
SDP
Catalog No. 5090108677
Encompass
CAS# 10043-35-3. 10g. Boric acid (99.9995%-B) PURATREM. MFCD00011337. Molecular Weight: 61.83. Molecular Formula: H3BO3. Color/form: white xtl. Strem# 05-0205. http://www.strem.com/catalog/v/05-0205/
Catalog No. 50-901-08677 Supplier Strem, An Ascensus Company Supplier No. 05020510G
CAS# 10043-35-3. 10g. Boric acid (99.9995%-B) PURATREM. MFCD00011337. Molecular Weight: 61.83. Molecular Formula: H3BO3. Color/form: white xtl. Strem# 05-0205. http://www.strem.com/catalog/v/05-0205/