Learn More
Alkali Scientific Boric Acid, Crystals, ACS Grade, 5 Kilograms
SDP
Catalog No. 5000660306
Encompass_Preferred
Boric Acid (5 kg, ACS Grade) is a high-purity crystalline chemical compound used in laboratory and industrial applications. As an acid, boric acid is commonly used in the manufacture of buffer solutions, and as an antiseptic in pharmaceuticals and cosmetics. It is also used in various chemical syntheses, such as the production of borosilicate glass, fiberglass, and flame retardants. Boric acid’s antimicrobial properties make it valuable in disinfectants and personal care products. It is also employed in agricultural applications as a micronutrient for plants. This ACS grade boric acid meets strict quality standards, ensuring consistency and reliability in laboratory testing.
Catalog No. 50-006-60306 Supplier Alkali Scientific Supplier No. BOR005CP
Boric Acid (5 kg, ACS Grade) is a high-purity crystalline chemical compound used in laboratory and industrial applications. As an acid, boric acid is commonly used in the manufacture of buffer solutions, and as an antiseptic in pharmaceuticals and cosmetics. It is also used in various chemical syntheses, such as the production of borosilicate glass, fiberglass, and flame retardants. Boric acid’s antimicrobial properties make it valuable in disinfectants and personal care products. It is also employed in agricultural applications as a micronutrient for plants. This ACS grade boric acid meets strict quality standards, ensuring consistency and reliability in laboratory testing.