Learn More
Alkali Scientific Boric Acid, Crystals, ACS Grade, 500 Grams
SDP
Catalog No. 5000660755
Encompass_Preferred
Boric Acid, ACS Grade, in crystalline form is commonly used in laboratory applications, particularly for buffer preparation and chemical synthesis. The 500g packaging provides a manageable quantity suitable for laboratories working with smaller batches. Boric acid serves as a pH buffer in various chemical reactions, particularly in biological and pharmaceutical research. Its ability to stabilize pH in solutions makes it valuable in the preparation of buffers for enzyme studies, nucleic acid work, and other sensitive biochemical assays. This high-purity boric acid meets ACS standards, ensuring it is suitable for analytical and experimental purposes requiring exacting chemical consistency.
Catalog No. 50-006-60755 Supplier Alkali Scientific Supplier No. BOR001CP
Boric Acid, ACS Grade, in crystalline form is commonly used in laboratory applications, particularly for buffer preparation and chemical synthesis. The 500g packaging provides a manageable quantity suitable for laboratories working with smaller batches. Boric acid serves as a pH buffer in various chemical reactions, particularly in biological and pharmaceutical research. Its ability to stabilize pH in solutions makes it valuable in the preparation of buffers for enzyme studies, nucleic acid work, and other sensitive biochemical assays. This high-purity boric acid meets ACS standards, ensuring it is suitable for analytical and experimental purposes requiring exacting chemical consistency.