Learn More
Sigma Aldrich Fine Chemicals Biosciences Boric acid puriss. p.a. AC500G
Catalog No. 502898958
Encompass
Boric acid is a very weak acid having pH in the range of 3.6-4.0. Coulometric method for the determination of boric acid has been reported.
Catalog No. 50-289-8958 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 31146500G
Boric acid is a very weak acid having pH in the range of 3.6-4.0. Coulometric method for the determination of boric acid has been reported.