Learn More
Sigma Aldrich Fine Chemicals Biosciences Boric acid puriss pa AC25KG
Catalog No. 502898303
Encompass
Boric acid is a very weak acid having pH in the range of 3.6-4.0. Coulometric method for the determination of boric acid has been reported.
Catalog No. 50-289-8303 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 311462.5KG
Boric acid is a very weak acid having pH in the range of 3.6-4.0. Coulometric method for the determination of boric acid has been reported.