Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Boric Acid, Reagent, Powder, A.C.S. 500 GR
SDP

Catalog No. 50980211
Encompass_Preferred

CAS #10043-35-3 H₃BO₃ F.W. 61.83
Assay: 99.95%

A buffer component in Tris-EDTA-Borate buffer. Also used in Silver-Methenamine buffer. Swift, J.A. (1968). The Electron histochemistry of cystine-containing proteins in thin transverse sections of human hair. J.R. Microsc. Soc. 88,449.

Catalog No. 50-980-211 Supplier Electron Microscopy Sciences Supplier No. 11460
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS #10043-35-3 H₃BO₃ F.W. 61.83
Assay: 99.95%

A buffer component in Tris-EDTA-Borate buffer. Also used in Silver-Methenamine buffer. Swift, J.A. (1968). The Electron histochemistry of cystine-containing proteins in thin transverse sections of human hair. J.R. Microsc. Soc. 88,449.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.