Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI Boric Acid with Sher Indicator, pH 4.65
SDP

Catalog No. 05405672 Shop All BUCHI Products
Change view
Click to view available options
Concentration or Composition (by Analyte or Components):
0.02
0.04
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration or Composition (by Analyte or Components)
05-405-672 0.02
05-405-673 0.04
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05-405-672 Supplier BUCHI Supplier No. 11064972
Add to Cart
Add to Cart

Boric Acid with Sher Indicator, pH adjusted to 4.65 ± 0.15 for pH or colorimetric titration.

  • Ready to use boric acid
  • Maintains the correct concentration, pH, and indicator ratio
  • For pH or colorimetric titration
  • pH adjusted to 4.65 ± 0.15
  • For pH or colorimetric titration

Specifications

Chemical Name or Material Boric Acid with Sher Indicator
CAS 64-17-5, 1310-73-2, 10043-35-3, 7732-18-5, 76-60-8
Quantity 5 L
Molecular Formula BH3O3
Packaging Plastic Bottle
Synonym Orthoboric Acid
Molecular Weight (g/mol) 61.83
Concentration or Composition (by Analyte or Components) 0.02
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.