Learn More
Hach Company Boric Acid Solution (Indicating), 20 g/L, 1 L
Catalog No. 501995828 Shop All Hach Company Products
Encompass
For determination of ammonia by the phenate method. 20 g/L H3BO3, with indicators, APHA formulation 1 liter bottle
Catalog No. 50-199-5828 Supplier Hach Company Supplier No. 2339053
For determination of ammonia by the phenate method. 20 g/L H3BO3, with indicators, APHA formulation 1 liter bottle