Learn More
Bioworld Boric Acid, 2% (w/v), 4 L
Catalog No. 50255494
Encompass_Preferred
Boric Acid Solution 2% (w/v) in Biotechnology Grade H2OA commonly used molecular biology reagentAppearance: LiquidBoiling Point : 300 °CDensity : 1.44 g/cm3Melting Point: 168.9 °C (dec.)RTECS Number: ED4550000
Catalog No. 50-255-494 Supplier Bioworld Supplier No. 402201023
Boric Acid Solution 2% (w/v) in Biotechnology Grade H2OA commonly used molecular biology reagentAppearance: LiquidBoiling Point : 300 °CDensity : 1.44 g/cm3Melting Point: 168.9 °C (dec.)RTECS Number: ED4550000