Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Acid Specimen Container with 1 g Tablet, Cenmed™
SDP

Catalog No. 21520247
Change view
Click to view available options
Quantity:
100 Containers, 1 x 1 g Tablet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-520-247 100 Containers, 1 x 1 g Tablet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-520-247 Supplier Cen-Med Enterprises Supplier No. CENMED 3250205
Add to Cart
Add to Cart
This item is not returnable. View return policy

Preserve your samples effectively with the Boric Acid Specimen Container with 1 g tablet. This non-sterile container is designed for optimal sample preservation, ensuring the integrity of your specimens.

  • Non-sterile container is crafted for use in non-sterile environments, providing a reliable solution for sample storage in various settings
  • Ensure the integrity of your specimens with a container specifically designed for optimal sample preservation, maintaining the quality of your samples over time
  • Included 1 g tablet of boric acid enhances the container's preservation capabilities, making it an ideal choice for various sample types
  • Reliable and convenient solution
  • Elevate your sample storage with a solution that ensures ideal sample quality, preserving the characteristics of your specimens for accurate analysis

Specifications

Physical Form Tablets, Liquid
CAS 10043-35-3
Color Clear
Quantity 100 Containers, 1 x 1 g Tablet
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.