Learn More
National Diagnostics BORIC ACID - ULTA PURE 500 GR
SDP
Catalog No. 5089990052
Encompass
Boric Acid - ULTA PURE, Primary component of TBE buffer, the most commonly used electrophoresis buffer for DNA and RNA, 500 GR
Catalog No. 50-899-90052 Supplier National Diagnostics Supplier No. EC609500GRM
Boric Acid - ULTA PURE, Primary component of TBE buffer, the most commonly used electrophoresis buffer for DNA and RNA, 500 GR