Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Anhydride (Laboratory), Fisher Chemical

Catalog No. A763 Shop All Fisherbrand Products
Change view
Click to view available options
Glass Bottle; 3kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
A76-3 Glass Bottle; 3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A76-3

Specifications

pH 4
Color White
Melting Point 450°C
CAS 1303-86-2
Boiling Point 1860°C
Density 2.460 g/cm3
Identification Pass Test
Packaging Glass Bottle
Assay Percent Range ≥98 %
Quantity 3 kg
Molecular Formula B2O3
Physical Form Solid
MDL Number MFCD00011315
Synonym boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g
InChI Key JKWMSGQKBLHBQQ-UHFFFAOYSA-N
SMILES O=BOB=O
IUPAC Name oxo[(oxoboranyl)oxy]borane
Molecular Weight (g/mol) 69.62
PubChem CID 518682
ChEBI CHEBI:30163
Formula Weight 69.62
Percent Purity ≥98%
Grade Laboratory
Chemical Name or Material Boric Anhydride
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
May impair fertility. May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Obtain medical attention. Do not induce vomiting.

NFPA

Health:2
Flammability:0
Instability:1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.