Learn More
Grainger Micro 100 Boring Bar, Carbide, TiN, 3/16 in. Shank, 1 ea.
Catalog No. 502756279
Encompass_Preferred
This boring bar is engineered for precision boring applications. Constructed from solid carbide for enhanced durability and performance, it features a TiN coating to improve wear resistance in various machining environments.
  • Carbide construction provides rigidity and extended tool life.
  • TiN coating facilitates reduced friction and improved surface finish.
  • Right hand cutting direction suitable for standard boring operations.
  • Features corner radius profile and split geometry for effective chip control.
  • Straight shank with three flats for secure clamping.
  • Maximum bore depth of 0.6000 in.
  • Minimum bore diameter of 0.1400 in.
Catalog No. 50-275-6279 Supplier Grainger Supplier No. 15R882
This boring bar is engineered for precision boring applications. Constructed from solid carbide for enhanced durability and performance, it features a TiN coating to improve wear resistance in various machining environments.
  • Carbide construction provides rigidity and extended tool life.
  • TiN coating facilitates reduced friction and improved surface finish.
  • Right hand cutting direction suitable for standard boring operations.
  • Features corner radius profile and split geometry for effective chip control.
  • Straight shank with three flats for secure clamping.
  • Maximum bore depth of 0.6000 in.
  • Minimum bore diameter of 0.1400 in.