Learn More
Apexbio Technology LLC Borneol,1gCAS# 507-70-0
SDP
Supplier:  Apexbio Technology LLC N14931000
Encompass_Preferred
Extracted from Cinnamomum cassia;Suitability:Ethanol,ether,petroleum ether,benzene,toluene,acetone,decalin,tetralin;Store the product in sealed,cool and dry condition. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0513