Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Borneol, ≥97.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018356 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8356 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8356 Supplier MilliporeSigma Supelco Supplier No. 6887810MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Borneol, a bicyclic monoterpenoid alcohol which is a major constituent of medicinal plants is grouped under the volatile organic compounds (VOCs) family of chemicals. It is employed as analgesic and anesthetic in traditional Chinese and Japanese medicines.

Featured Industries: Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 464-43-7
Flash Point 65°C - closed cup
Physical Form Neat
Linear Formula C10H18 O
MDL Number MFCD00066427
Melting Point 206°C to 209°C
Synonym endo-(1 R)-1,7,7-Trimethylbicyclo[2.2.1 ]heptan-2-ol
Type Analytical Standard
Quantity 10 mg
Formula Weight 154.25
Percent Purity ≥97.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.