Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borneol (contains ca. 20% Isoborneol) 70.0+%, TCI America™
SDP

Catalog No. B0525500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0525500G 500 g
B052525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0525500G Supplier TCI America Supplier No. B0525500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 507-70-0
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00066426
InChI Key DTGKSKDOIYIVQL-HHCGNCNQSA-N
PubChem CID 44630107
IUPAC Name (1R,3S)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-ol
SMILES CC1(C2CCC1(C(C2)O)C)C

Specifications

Melting Point 201°C
Color White
Quantity 500 g
UN Number 1312
Formula Weight 154.25
Percent Purity ≥70.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Borneol (contains ca. 20% Isoborneol)
EINECSNumber (4)-0607
RTECSNumber ED7000000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.