Learn More
Medchemexpress LLC (+)-Bornyl acetate | 20347-65-3 | MFCD00135942 | 98.0% | 196.29 g·mol⁻¹ | C12H20O2 | 10 MG
SDP
Catalog No. 5000297771
Encompass_Preferred
(+)-Bornyl acetate is a naturally occurring monoterpene ester found in pichtae (Siberian fir needle) essential oil. Supplied as a research-grade reagent with reported biological activity including inhibition of Arabidopsis root growth, it is used in aroma, flavor, and plant physiology studies. Typical properties include CAS 20347-65-3, molecular formula C12H20O2, molecular weight 196.29 g·mol⁻¹, and reported purity 98%.
  • Natural monoterpene ester found in fir needle essential oil.
  • Reported to inhibit root growth in Arabidopsis seedlings.
  • High purity suitable for research applications (98%).
  • Supplied in small, precise quantities appropriate for analytical and biological assays.
  • Volatile aroma compound with applications in flavor and fragrance research.
  • Applicable to plant physiology and bioactivity studies.
Catalog No. 50-002-97771 Supplier Medchemexpress LLC Supplier No. HYB191810MG
(+)-Bornyl acetate is a naturally occurring monoterpene ester found in pichtae (Siberian fir needle) essential oil. Supplied as a research-grade reagent with reported biological activity including inhibition of Arabidopsis root growth, it is used in aroma, flavor, and plant physiology studies. Typical properties include CAS 20347-65-3, molecular formula C12H20O2, molecular weight 196.29 g·mol⁻¹, and reported purity 98%.
  • Natural monoterpene ester found in fir needle essential oil.
  • Reported to inhibit root growth in Arabidopsis seedlings.
  • High purity suitable for research applications (98%).
  • Supplied in small, precise quantities appropriate for analytical and biological assays.
  • Volatile aroma compound with applications in flavor and fragrance research.
  • Applicable to plant physiology and bioactivity studies.