Learn More
Medchemexpress LLC Bornyl acetate | 76-49-3 | 196.29 | 1 ML
SDP
Catalog No. 5000417121
Encompass_Preferred
Bornyl acetate is a potent odorant and possesses anti-cancer activity. It induces dose-dependent growth inhibitory effects on human gastric cancer cells in vitro and enhances cell death via apoptosis when combined with 5-FU. This liquid compound is stable under recommended storage conditions, with various storage guidelines for both neat and in-solvent forms.
  • Potent odorant with high flavor dilution factor
  • Exhibits anti-cancer activity in human gastric cancer cells
  • Induces dose-dependent growth inhibition and apoptosis
  • Available as a 10 mM solution in DMSO
  • Recommended storage at 4°C, sealed and away from moisture
  • In-solvent storage at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-17121 Supplier Medchemexpress LLC Supplier No. HYN075610MM/1ML
Bornyl acetate is a potent odorant and possesses anti-cancer activity. It induces dose-dependent growth inhibitory effects on human gastric cancer cells in vitro and enhances cell death via apoptosis when combined with 5-FU. This liquid compound is stable under recommended storage conditions, with various storage guidelines for both neat and in-solvent forms.
  • Potent odorant with high flavor dilution factor
  • Exhibits anti-cancer activity in human gastric cancer cells
  • Induces dose-dependent growth inhibition and apoptosis
  • Available as a 10 mM solution in DMSO
  • Recommended storage at 4°C, sealed and away from moisture
  • In-solvent storage at -80°C for 6 months or -20°C for 1 month