Learn More
Medchemexpress LLC Bornyl acetate | 76-49-3 | 196.29 | 500 MG
SDP
Catalog No. 5000417137
Encompass_Preferred
This product is found in pichtae essential oil (Siberian fir needle oil). It has a stronger inhibitory effect on root growth of *Arabidopsis* seedlings.
  • Found in pichtae essential oil.
  • Inhibits root growth in *Arabidopsis* seedlings.
Catalog No. 50-004-17137 Supplier Medchemexpress LLC Supplier No. HYN0756500MG
This product is found in pichtae essential oil (Siberian fir needle oil). It has a stronger inhibitory effect on root growth of *Arabidopsis* seedlings.
  • Found in pichtae essential oil.
  • Inhibits root growth in *Arabidopsis* seedlings.