Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (-)-Bornyl Acetate

Catalog No. B552035G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B552035G 5 g
B55203100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B552035G Supplier Sigma Aldrich Supplier No. B552035G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(-)-Bornyl acetate can be used:As a key intermediate in the preparation of camphor.To prepare (-)-englerin A, an anti-cancer agent.

Specifications

CAS 79-08-3
Melting Point 47°C to 49°C
Boiling Point 208°C
Molecular Formula C2H3BrO2
Linear Formula BrCH2COOH
Refractive Index n20/D 1.463
MDL Number MFCD00002678
Quantity 5 g
Synonym endo-(1S)-1,7,7-Trimethylbicyclo[2.2.1]hept-2-yl acetate
Molecular Weight (g/mol) 138.95 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.