Learn More
Foxx Life Sciences Borosil® Pear-Shaped Glass Sepratery Funnel with Boroflo Stopcock PTFE key, Interchangeable Glass Stopper (24/29), 250 mL, 10/CS
Supplier:  Foxx Life Sciences 6402021
Encompass_Preferred
Borosil® Separatory Funnels use a self-lubricating, replaceable PTFE stopcock that does not require grease or lubrication in order to achieve fine, smooth control of flow. Glass pennyhead stoppers included. Borosil® Boroflo Sepratery Funnels has best performance in the world. Borosil® Separatory Funnels comply with DIN/ISO 4800. 60mL and 125mL options are not covered in DIN/ISO. Available from 60mL to 2L. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices. And thanks to a unique annealing process, Borosil® Glassware is stress-free and incredibly resistant to thermal shock.
Catalog No. 50-172-0537