Learn More
AdipoGen Boromycin
SDP
Supplier:  AdipoGen AGCN20166C250
Encompass_Preferred
Chemical. CAS 34524-20-4. Formula C45H74BNO15. MW 879.9. Isolated from Streptomyces sp. Boron-containing macrolide antibiotic. Antibacterial. Antiprotozoal Plasmodium, Babesia. Anti-HIV compound. Ca2 homeostasis modulator. Potentiator of bleomycin-induced anti-tumor activity. Used as a chiral selector in capillary electrophoresis.
Catalog No. 50-256-2648