Learn More
AdipoGen Boromycin (1 mg)
SDP
Supplier:  AdipoGen AGCN20166M001
Encompass_Preferred
Chemical. CAS: 34524-20-4. Formula: C45H74BNO15. MW: 879.9. Isolated from Streptomyces sp. Boron-containing macrolide antibiotic. Antibacterial. Antiprotozoal (Plasmodium, Babesia). Anti-HIV compound. Ca2+ homeostasis modulator. Potentiator of bleomycin-induced anti-tumor activity. Used as a chiral selector in capillary electrophoresis.
Catalog No. 50-205-3557