Learn More
Medchemexpress LLC Boron, [1-[3-[5-[(3,5-dimethyl-2H-pyrrol-2-ylidene-KN)methyl]-1H-pyrrol-2-yl-KN]-1-oxopropoxy]- | 146616-66-2 | 95.60% | 389.16 | 100 MG
SDP
Catalog No. 5000344042
Encompass_Preferred
BODIPY FL NHS ester (BODIPY FL, SE) is a cell membranes-penetrable amine-reactive fluorescent probe. The maximum excitation/emission wavelength of this compound are 502/511 nm, respectively. It has high stability and is insensitive to the polarity, pH and type of solvent, and can maintain stable fluorescence properties under different environmental conditions. It can be used for the synthesis of protease substrates, live cell imaging, protein labeling and immunoassay.
  • Cell membranes-penetrable amine-reactive fluorescent probe
  • Maximum excitation/emission wavelength are 502/511 nm
  • High stability and insensitive to polarity, pH, and solvent type
  • Maintains stable fluorescence properties under different environmental conditions
  • Can be used for synthesis of protease substrates
  • Suitable for live cell imaging
  • Useful for protein labeling
  • Applied in immunoassay
Catalog No. 50-003-44042 Supplier Medchemexpress LLC Supplier No. HY11021050MG
BODIPY FL NHS ester (BODIPY FL, SE) is a cell membranes-penetrable amine-reactive fluorescent probe. The maximum excitation/emission wavelength of this compound are 502/511 nm, respectively. It has high stability and is insensitive to the polarity, pH and type of solvent, and can maintain stable fluorescence properties under different environmental conditions. It can be used for the synthesis of protease substrates, live cell imaging, protein labeling and immunoassay.
  • Cell membranes-penetrable amine-reactive fluorescent probe
  • Maximum excitation/emission wavelength are 502/511 nm
  • High stability and insensitive to polarity, pH, and solvent type
  • Maintains stable fluorescence properties under different environmental conditions
  • Can be used for synthesis of protease substrates
  • Suitable for live cell imaging
  • Useful for protein labeling
  • Applied in immunoassay