Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Boron

Catalog No. 2666205G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-662-05G 5 g
26-662-025G -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 26-662-05G Supplier Sigma Aldrich Supplier No. 2666205G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Boron can be used as a: Dopant on single-walled carbon nanotubes (SWCNT) used as a catalyst for acetylene hydrochlorination.Catalyst for enhancing the graphitization of various industrial carbons.

Specifications

CAS 7440-42-8
Density 2.34 g/mL at 25°C
Quantity 5 g
Molecular Formula B
Linear Formula B
MDL Number MFCD00134034
Molecular Weight (g/mol) 10.81 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.