Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Boron

Catalog No. 1558050GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-580-50GF 50 g
15-580-250GF 250 g
15-580-5GF 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Warning
Catalog No. 15-580-50GF Supplier Sigma Aldrich Supplier No. 1558050GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Boron (amorphous powder) has been used as a boron source to synthesize hexagonal boron nitride (h-BN), boron doped diamond (BDD) and europium-doped BN nanotubes.

Specifications

CAS 7440-42-8
Density 2.34 g/mL at 25°C
Quantity 50 g
Molecular Formula B
Linear Formula B
MDL Number MFCD00134034
Molecular Weight (g/mol) 10.81 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.