Learn More
Electron Microscopy Sciences Boron Carbide Powder, 50 µm particles 1 LB
SDP
Catalog No. 50997435
Encompass_Preferred
Boron Carbide is one of the hardest materials known, ranking third behind diamond and cubic boron nitride. This makes it is ideal for use as an abrasive in polishing and lapping applications. Extreme hardness (Knoop 100g: 2900-3580 kg/mm2), Good chemical resistance, Good nuclear properties, Low density (2.52 g/cm2)
Particles: 50 µm
Catalog No. 50-997-435 Supplier Electron Microscopy Sciences Supplier No. 5051050
Boron Carbide is one of the hardest materials known, ranking third behind diamond and cubic boron nitride. This makes it is ideal for use as an abrasive in polishing and lapping applications. Extreme hardness (Knoop 100g: 2900-3580 kg/mm2), Good chemical resistance, Good nuclear properties, Low density (2.52 g/cm2)
Particles: 50 µm