Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Boron nitride

Catalog No. 79053210G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-053-210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 79-053-210G Supplier Sigma Aldrich Supplier No. 79053210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Boron nitride has been well studied for applications including ceramic and polymer nanocomposites. Nanoscale boron nitride is essential for applications at this length scale as the increased surface area resulting in better incorporation in the matrix. BN possesses a number of material properties which increase its utility in machining and coating applications including wear resistance; chemical and thermal stability and wide band gap.

Specifications

CAS 10043-11-5
Quantity 10 g
Molecular Formula BN
Linear Formula BN
MDL Number MFCD00011317
Molecular Weight (g/mol) 24.82
Percent Purity 99% trace metals basis
Density 2.29 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.