Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Boron nitride

Catalog No. 25547550G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-547-550G 50 g
25-547-5250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25-547-550G Supplier Sigma Aldrich Supplier No. 25547550G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hexagonal boron nitride (hBN) has a layered structure similar to graphite and can be exfoliated as singlelayered BN sheets. hBN has applications in catalysts, optoelectronics and semiconductor devices. Boron nitride posseses high thermal conductivity of approximately 400W/mK at 300K.Hexagonal boron nitride powder has been used:In the fabrication and luminescence study of monolayered boron nitride quantum dots.Rare earth boron complexes.Boron nitride filled polybenzoxazine system as a thermally conducting molding components for electronic packaging applications. Boron containing ceramic products. As a starting material to form BN nanotubes. To load silicone rubber and its consecutive effect on its thermal, mechanical and morphological properties was studied.

Specifications

CAS 10043-11-5
Quantity 50 g
Molecular Formula BN
Linear Formula BN
MDL Number MFCD00011317
Molecular Weight (g/mol) 24.82
Percent Purity 98%
Density 2.29 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.