Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boron nitride, 99.5% (metals basis), Thermo Scientific™

Catalog No. AA44834KJ
Change view
Click to view available options
Quantity:
5 pc.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA44834KJ 5 pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA44834KJ Supplier Thermo Scientific Chemicals Supplier No. 44834KJ

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 10043-11-5
Quantity 5 pc.
Molecular Formula BN
MDL Number MFCD00011317
Synonym boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
InChI Key PZNSFCLAULLKQX-UHFFFAOYSA-N
SMILES B#N
IUPAC Name boranylidyneamine
Molecular Weight (g/mol) 24.82
PubChem CID 66227
ChEBI CHEBI:50883
Formula Weight 24.82
Percent Purity 99.5%
Packaging Bag
Chemical Name or Material Boron nitride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.