Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boron nitride, Refractory Brushable Paint, BN 31%

Catalog No. p-4824632
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4408618 50 g
AA4408630 250 g
AA44086A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4408618 Supplier Thermo Scientific Chemicals Supplier No. 04408618
Only null left
Add to Cart

CAS: 10043-11-5 | BN | 24.82 g/mol

Boron nitride is used as a lubricant and an additive to cosmetic products. It is also used in parts of high-temperature equipment, nanotechnology, polymorph. It is also used in used in xerographic process and laser printers. It is also used in fiberglass, ceramics, wood preservative, nuclear application and pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-11-5
Molecular Formula BN
Molecular Weight (g/mol) 24.82
MDL Number MFCD00011317
InChI Key LNLSXDSWJBUPHM-UHFFFAOYSA-N
Synonym boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
PubChem CID 66227
ChEBI CHEBI:50883
SMILES B=N

Specifications

Physical Form Paste
Quantity 50 g
Solubility Information Soluble in water (slightly soluble) at 20°C,and water (soluble) at 95°C.
IUPAC Name boranylidyneamine
Chemical Name or Material Boron nitride
Hazard Category H301-H332: Toxic if swallowed. Harmful if inhaled.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.